missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17B Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1709-SP
This item is not returnable.
View return policy
Description
IL-17B Polyclonal specifically detects IL-17B in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-17B | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27190 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Blockade of Receptor-ligand Interaction, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q9QXT6 | |
| IL17B | |
| E. coli-derived recombinant mouse IL-17B His21-Phe180 Accession # Q9QXT6 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-17B in ELISAs and Western blots. In sandwich immunoassays, approximately 40% cross-reactivity with recombinant human IL-17B is observed and less than 0.3% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17C, rmIL-17D, rmIL-17E, and rmIL-17F is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction