missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17E/IL-25 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC13991S-100UG
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_542767 | |
| IL25 | |
| E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767 | |
| RUO | |
| 64806 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 207702 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-17E/IL-25 in ELISAs. In ELISAs, no cross-reactivity with recombinant human IL-17E, recombinant mouse (rm) IL-17, rmIL-17B, C, D, or F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction