missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-17E/IL-25 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18716177
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18716177 25 μg 25µg
18071522 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18716177 Supplier R&D Systems Supplier No. AF1399SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IL-17E/IL-25 Polyclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17E/IL-25
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_542767
Gene Alias IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Gene Symbols IL25
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64806
Test Specificity Detects mouse IL-17E/IL-25 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17E and recombinant rat IL-17E is observed, and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rhIL-17C, rhIL-17D, and rhIL-17F is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.