missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17RA/IL-17R Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB448
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for Western Blot.
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23765 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 105828 | |
| Western Blot 2 ug/mL | |
| Q60943 | |
| IL17RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-17 RA/IL-17 R Extracellular domain Accession # Q60943 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-17 RA/IL-17 R in direct ELISAs and Western blots. Shows 50-100% cross-reactivity with recombinant human IL-17 RA/IL-17 R and no cross-reactivity with recombinant mouse IL-17B R. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction