missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-17RA/IL-17R Antibody, R&D Systems™

Product Code. 18038601
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18038601 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18038601 Supplier R&D Systems Supplier No. AF448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 8 publications

IL-17RA/IL-17R Polyclonal specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17RA/IL-17R
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Alias CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Gene Symbols IL17RA
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-17 RA/IL-17 R Extracellular domain
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Test Specificity Detects mouse IL-17 RA/IL-17 R in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human IL-17 RA/IL-17 R is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-17 RC and rmIL-17 RD is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.