missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-18 BPc Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF129
This item is not returnable.
View return policy
Description
IL-18 BPc Polyclonal specifically detects IL-18 BPc in Mouse samples. It is validated for Western Blot.
Specifications
| IL-18 BPc | |
| Polyclonal | |
| Unconjugated | |
| AAD17193 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 16182 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Il18r1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-18 BPc Arg25-Ser192 Accession # AAD17193 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-18 BPc in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse IL-18 BPd is observed and less than 1% cross-reactivity with recombinant human IL-18 BPa is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction