missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-18 BPd Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF122
This item is not returnable.
View return policy
Description
IL-18 BPd Polyclonal specifically detects IL-18 BPd in Mouse samples. It is validated for Western Blot, ELISA, Neutralization.
Specifications
| IL-18 BPd | |
| Polyclonal | |
| Unconjugated | |
| AAD17194 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 16068 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL | |
| Il18bp | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 BPd Thr27-Ala191 Accession # AAD17194 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-18 BPd in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction