missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1ra/IL-1F3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB4801
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Mouse samples. It is validated for Western Blot.
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q542W1 | |
| IL1RN | |
| E. coli-derived recombinant mouse IL-1ra/IL-1F3 Arg8-Gln159 Accession # Q542W1 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-1ra/IL-1F3 in direct ELISAs and Western blots. In Western blots, approximately 30%-50% cross-reactivity with recombinant equine IL-1ra and recombinant human (rh) IL-1ra is observed. Less than 5% cross-reactivity with recombinant mouse (rm) IL-1b, rmIL-18, rmIL-36Ra, rmIL-36a, rmIL-36b, rmIL-1F10, rhIL-36, rhIL-1F7, or recombinant porcine IL-1ra, or recombinant rat IL-1ra is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 694204 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3557 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction