missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC582RS-100UG
This item is not returnable.
View return policy
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-22 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession # Q9JJY9 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 140301R | |
| Intracellular Staining by Flow Cytometry | |
| Q9JJY9 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 50616 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction