missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-24 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2786-SP
This item is not returnable.
View return policy
Description
IL-24 Monoclonal specifically detects IL-24 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IL-24 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_444325 | |
| IL24 | |
| E. coli-derived recombinant mouse IL-24 Arg27-His181 Accession # NP_444325 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-24 in direct ELISAs and Western blots. No cross-reactivity with recombinant human IL-24 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 303308 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C49A, FISP, IL10B, IL24, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, MOB-5, ST16, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11009 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction