missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-27 p28 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC1834N-100UG
This item is not returnable.
View return policy
Description
IL-27 Monoclonal specifically detects IL-27 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-27 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O35228 (EBI3) & Q8K3I6 (p28) | |
| Il27 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27 Tyr19-Pro228 (EBI3) & Phe29-Ser234 (p28) Accession # O35228 (EBI3) & Q8K3I6 (p28) | |
| RUO | |
| 246778 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 234205 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL-27, IL27 p28, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, IL-30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-27 in Western blots. This antibody recognizes the p28 subunit either as part of a p28/EBI-3 heterodimer or as a free subunit. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction