missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-27 p28/IL-30 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7430-SP
This item is not returnable.
View return policy
Description
IL-27 p28/IL-30 Monoclonal specifically detects IL-27 p28/IL-30 in Mouse samples. It is validated for Western Blot.
Specifications
| IL-27 p28/IL-30 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8K3I6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 246779 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 744015 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Il27 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27 p28/IL-30 Phe29-Ser234 Accession # Q8K3I6 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-27 p28/IL-30 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-27/IL-35 EBI3 subunit, recombinant human (rh) IL-27/IL-35 EBI3 subunit, or rhIL-27 heterodimer is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction