missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-28B/IFN-lambda 3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1789
This item is not returnable.
View return policy
Description
IL-28B/IFN-lambda 3 Monoclonal specifically detects IL-28B/IFN-lambda 3 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-28B/IFN-lambda 3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1.5-6.0 ug/mL | |
| Q8CGK6 | |
| Ifnl3 | |
| E. coli-derived recombinant mouse IL-28B/IFN-lambda 3 Asp20-Val193 Accession # Q8CGK6 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-28B/IFN-lambda 3 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse (rm) IL-28A/IFN-gamma 2 and no cross-reactivity with recombinant human (rh) IL-28A or rhIL-29 is observed. This antibody neutralizes 100% of the bioactivity of rmIL-28A/IFN-gamma 2. | |
| Mouse | |
| Purified |
| Western Blot, Neutralization | |
| 244710 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| interleukin-28B, IFN-lambda 3, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 282617 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction