missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse IL-3 R alpha / CD123 Protein, His Tag

Product Code. 16437739
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437739 1 mg 1mg
16427739 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437739 Supplier ACROBiosystems Supplier No. ILAM52H41mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SPR verified

Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.4 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_032395.1
Molecular Weight (g/mol) 36.4 kDa
Quantity 1 mg
Source Mouse IL-3 R alpha, His Tag (ILA-M52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 17- Lys 331 (Accession # P26952-1).
Immunogen Ser 17- Lys 331
Storage Requirements -20°C
Gene Alias IL3R,IL3RA,IL-3 Ra,IL-3 R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3 Ra,MGC34174,IL-3 R alpha
Common Name IL-3 R alpha
Conjugate Unconjugated
Species Mouse
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.