missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-4 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB404R-500
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-4 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| E.coli-derived recombinant mouse IL-4 His23-Ser140 Accession No. P07750 | |
| 500 ug | |
| Primary | |
| Detects mouse IL-4 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ELISA, Neutralization | |
| 30340R | |
| Neutralization 0.1-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07750 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3565 | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction