Learn More
Mouse IL-6R alpha Biotinylated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems BAM1830
Product Code. 18180471
  • € 510.00 / 100µg
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6R alpha
Monoclonal
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
P22272
Il6ra
Mouse myeloma cell line NS0-derived recombinant mouse IL-6 R alpha Leu20-Glu357 Accession # P22272
100 μg
Primary
Detects mouse IL-6 R alpha .
Mouse
Purified
Flow Cytometry, Immunocytochemistry
255821
Biotin
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
3570
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.