missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Mouse Il31 (Q6EAL8) Recombinant Protein

Product Code. 16101650
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16101650 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16101650 Supplier Abnova™ Supplier No. P4632.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: MTCSLSFGAPISKEDLRTTIDLLKQESQDLYNNYSIKQASGMSADESIQLPCFSLDREALTNISVIIAHLEKVKVLSENTVDTSWVIRWLTNISCFNPLNLNISVPGNTDESYDCKVFVLTVLKQFSNCMAELQAKDNTTC

Specifications

Accession Number Q6EAL8
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 76399
Molecular Weight (g/mol) 15.7kDa
Name Il31 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
Quantity 10 μg
Immunogen MTCSLSFGAPISKEDLRTTIDLLKQESQDLYNNYSIKQASGMSADESIQLPCFSLDREALTNISVIIAHLEKVKVLSENTVDTSWVIRWLTNISCFNPLNLNISVPGNTDESYDCKVFVLTVLKQFSNCMAELQAKDNTTC
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias 1700013B14Rik
Common Name Il31
Gene Symbol Il31
Biological Activity The activity is determined by the dose dependent production of IL-8 by human PBMCs. The expected ED50 for this effect is 16-24pg/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.