missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 2/CD49b Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1740G-100UG
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q62469 | |
| ITGA2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 2/CD49b Tyr27-Thr1129 Accession # Q62469 | |
| 100 μg | |
| Primary | |
| Detects mouse Integrin alpha 2/CD49b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 3, alpha 4, alpha 5, alpha E, or alpha V is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 235033 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3673 | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction