missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Intelectin-1/Omentin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8074-SP
This item is not returnable.
View return policy
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Intelectin-1/Omentin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O88310 | |
| ITLN1 | |
| E. coli-derived recombinant mouse Intelectin-1/Omentin Ala20-Ser298 Accession # O88310 | |
| 25 μg | |
| Primary | |
| Detects mouse Intelectin-1/Omentin in ELISA. In direct ELISAs, no significant cross-reactivity with recombinant mouse Intelectin-2 and recombinant human Intelectin-1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 746420 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, Intelectin1, INTLHL1, ITLN1, Itlna, ITLNITLN-1, LFR, LFRIntestinal lactoferrin receptor, omentin, Omentin-1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55600 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction