missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IRF5 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB8447R-100UG
This item is not returnable.
View return policy
Description
IRF5 Monoclonal specifically detects IRF5 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IRF5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P56477 | |
| IRF5 | |
| E. coli-derived recombinant mouse IRF5 Ala208-Phe288 Accession # P56477 | |
| 100 μg | |
| Primary | |
| Detects mouse IRF5 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 903430 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3663 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction