missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kallikrein 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7928
This item is not returnable.
View return policy
Description
Kallikrein 1 Polyclonal specifically detects Kallikrein 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Kallikrein 1 | |
| Polyclonal | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| NP_081082, P15947 | |
| KLK1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein 5 precursor/Kallikrein 1 Val23-Arg67(Kallikrein 5 precursor), Ile25-Asp261(Kallikrein 1) Accession # NP_081082, P15947 | |
| 100 μg | |
| Primary | |
| Detects mouse Kallikrein 1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Kallikrein 1, recombinant mouse (rm) Kallikrein 5 and rmKallikrein 7 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, KLK1, Klk6, KLKR, Tissue kallikrein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3816 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction