missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Klre-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7099U-100UG
This item is not returnable.
View return policy
Description
Klre-1 Monoclonal specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Klre-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_705818 | |
| Klre1 | |
| Chinese hamster ovary cell line, CHO derived recombinant mouse Klre-1 Lys94-Lys226 Accession # NP_705818 | |
| RUO | |
| 3824 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 854929 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD94, Klre1, NKG2I | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Klre-1 in ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction