missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Klre-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7099-SP
This item is not returnable.
View return policy
Description
Klre-1 Polyclonal specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Klre-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD94, Klre1, NKG2I | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3824 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry, CyTOF | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_705818 | |
| Klre1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Klre-1 Lys94-Lys226 Accession # NP_705818 | |
| 25 μg | |
| Primary | |
| Detects mouse Klre-1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human NKG2D and recombinant mouse NKG2D is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction