missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse L1CAM Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5674V-100UG
This item is not returnable.
View return policy
Description
L1CAM Monoclonal specifically detects L1CAM in Mouse samples. It is validated for Flow Cytometry.
Specifications
| L1CAM | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse L1CAM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEPCAM, rhMCAM, rhNCAM, rhNCAM-L1, rhOBCAM, recombinant mouse (rm)MAdCAM-1, or rmOCAM is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 555 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| L1CAM | |
| Mouse cerebellum-derived partially purified L1CAM | |
| RUO | |
| 3897 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction