missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LAP (TGF-beta 1) Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7666N-100UG
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, transforming growth factor beta 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 1 Met1-Ser390 Accession # P04202 | |
| 100 μg | |
| Primary | |
| Detects mouse LAP (TGF-beta 1) in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 860206 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P04202 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction