missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LDLR Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2255G-100UG
This item is not returnable.
View return policy
Description
LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LDLR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6GTJ9 | |
| LDLR | |
| Mouse myeloma cell line NS0-derived recombinant mouse LDL R Ala22-Arg790 (Ala23Val, Cys27Gly) Accession # Q6GTJ9 | |
| 100 μg | |
| Primary | |
| Detects mouse LDL R in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LDL R or recombinant mouse LRP-6 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 263123 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3949 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction