missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LIFR alpha APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5990A
This item is not returnable.
View return policy
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LIFR alpha | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF R alpha, LIF R beta, LIF receptor, LIFR, LIF-R, LIFRa, SJS2, STWS, SWS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3977 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 673602 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P42703 | |
| LIFR | |
| Mouse myeloma cell line NS0-derived recombinant mouse LIF R alpha Leu44-Ser828 Accession # P42703 | |
| 100 Tests | |
| Primary | |
| Detects mouse LIF R alpha in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LIF R alpha is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction