missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LIGHT/TNFSF14 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB17943V-100UG
This item is not returnable.
View return policy
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LIGHT/TNFSF14 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
| Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14 Asp72-Val239 Accession # Q9QYH9 | |
| 100 μg | |
| 8740 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 906909 | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| Q9QYH9 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse LIGHT/TNFSF14 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction