missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Limitin/IFN-zeta Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB597
This item is not returnable.
View return policy
Description
Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot.
Specifications
| Limitin/IFN-zeta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6030405N23Rik, BGIF, Ifnz, IFN-z, IFNzeta, interferon zeta, Lmtn | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 319146 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 183727 | |
| Western Blot 1 ug/mL | |
| Q9R1T0 | |
| Ifnz | |
| Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182 Accession # Q9R1T0 | |
| 500 μg | |
| Primary | |
| Detects mouse Limitin/IFN-zeta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IFN-gamma is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction