Learn More
Mouse Lipin 3 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF4064
 View more versions of this product
Product Code. 18099161
  • € 436.00 / 100µg
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lipin 3
Polyclonal
Unconjugated
Q99PI4
LPIN3
E. coli-derived recombinant mouse Lipin 3 Thr428-Arg529 Accession # Q99PI4
100 μg
Primary
Detects mouse Lipin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Lipin 3 is observed and less than 5% cross-reactivity with rhLipin 1 and rhLipin 2 is observed.
Mouse
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, LPIN3, phosphatidate phosphatase LPIN3, SMP2
Goat
Affinity Purified
RUO
64900
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.