missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ly-6G (Gr-1) Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB10371U-100UG
This item is not returnable.
View return policy
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Ly-6G (Gr-1) | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| GR, Gr-1, Ly-6, Ly6G | |
| Ly-6G transfected EL-4J cell line, Accession # P35461 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1A8 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35461 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects mouse Ly-6G in Flow Cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction