missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LYVE-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems BAF2125
This item is not returnable.
View return policy
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LYVE-1 | |
| Polyclonal | |
| Biotin | |
| Q8BHC0 | |
| LYVE1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0 | |
| 50 μg | |
| Primary | |
| Detects mouse LYVE-1 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) LYVE-1 and less than 1% cross-reactivity with rhCD44 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10894 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction