missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse M-CSF R/CD115 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3818R-100UG
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| M-CSFR/CD115 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P09581 | |
| CSF1R | |
| Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R/CD115 Ala20-Ser511 Accession # P09581 | |
| RUO | |
| 1436 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 460615 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse M-CSF R/CD115 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human M-CSF R/CD115, recombinant mouse (rm) PDGF R beta , or rmFlt-3 Ligand is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction