missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MARCO Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB29561G-100UG
This item is not returnable.
View return policy
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry
Specifications
| MARCO | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2 | |
| Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 2359A | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| Q60754 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 8685 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction