missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Melanocortin-3 R/MC3R Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3737T-100UG
This item is not returnable.
View return policy
Description
Melanocortin-3 R/MC3R Monoclonal specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Melanocortin-3 R/MC3R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| BMIQ9, MC3, MC3R, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, Melanocortin3R, Melanocortin-3R, OB20, Obesity quantitative trait locus, OQTL | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects Melanocortin 3R/MC3R in direct ELISAs and Western blots. Stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 345404 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MC3R | |
| Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R | |
| RUO | |
| 4159 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction