Learn More
R&D Systems™ Mouse mGluR8 (NP_032200) VersaClone cDNA
Specifications
Specifications
| Accession Number | NP_032200 |
| Gene ID (Entrez) | 2918 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date |
| Species | Mouse |
| Quantity | 10 μg |