missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse MMP-10 ELISA Kit

Product Code. 17105659
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17105659

Brand: Invitrogen™ EM54RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Mouse MMP-10 quantitates mouse MMP-10 in serum, supernatant. The assay will exclusively recognize both natural and recombinant mouse MMP-10.

MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34 A; transin 2; Transin-2
Assay Range 2 to 1,000 pg/mL
Assay Sensitivity 2 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 17384
Gene Symbol MMP10
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MMP10
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.