missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MMR/CD206 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems FAB2535G
This item is not returnable.
View return policy
Description
MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Mouse samples. It is validated for Flow Cytometry.
Specifications
| MMR/CD206/Mannose Receptor | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4360 | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Q2HZ94 | |
| MRC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Ala1388 Accession # Q2HZ94 | |
| 100 Tests | |
| Primary | |
| Detects mouse MMR/CD206 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human MMR is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction