missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neogenin Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1079
This item is not returnable.
View return policy
Description
Neogenin Polyclonal specifically detects Neogenin in Mouse samples. It is validated for Western Blot.
Specifications
| Neogenin | |
| Polyclonal | |
| Biotin | |
| NP_032710 | |
| NEO1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 with a Asp442-Leu461 deletion Accession # NP_032710 | |
| 50 μg | |
| Primary | |
| Detects mouse Neogenin in Western blots. In this format, approximately 5% cross-reactivity with recombinant mouse DCC is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, NEO, NEO1, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGN, NGNDKFZp547A066 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4756 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction