missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Netrin-G2a Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2744-SP
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Netrin-G2a Monoclonal specifically detects Netrin-G2a in Mouse samples. It is validated for Western Blot.
Tekniske data
| Netrin-G2a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_598007 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84628 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 349719 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ntng2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007 | |
| 25 μg | |
| Primary | |
| Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-1, rmNetrin-4, rmNetrin-G1a, recombinant chicken (rch) Netrin-1, or rchNetrin-2 is observed. | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion