missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neuregulin-3/NRG3 EGF-like Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF753
This item is not returnable.
View return policy
Description
Neuregulin-3/NRG3 Polyclonal specifically detects Neuregulin-3/NRG3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neuregulin-3/NRG3 | |
| Polyclonal | |
| Unconjugated | |
| O35181 | |
| NRG3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NRG3 Glu284-Pro344 Accession # O35181 | |
| 100 μg | |
| Primary | |
| Detects mouse NRG3 EGF-like domain in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| HRG3, neuregulin 3, neuregulin 3 variant 1, neuregulin 3 variant 10, neuregulin 3 variant 11, neuregulin 3 variant 12, neuregulin 3 variant 13, neuregulin 3 variant 14, neuregulin 3 variant 3, neuregulin 3 variant 4, neuregulin 3 variant 5, neuregulin 3 variant 6, neuregulin 3 variant 7, neuregulin 3 variant 8, neuregulin 3 variant 9, Neuregulin3, neuregulin-3-like polypeptide, NRG3, pro-neuregulin-3, membrane-bound isoform, pro-NRG3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10718 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction