missing translation for 'onlineSavingsMsg'
Learn More

Mouse Neuropilin-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Product Code. 18775248
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18775248 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18775248 Supplier R&D Systems Supplier No. FAB5994V100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin-1
Applications Flow Cytometry
Classification Monoclonal
Clone 761705
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P97333
Gene Alias BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Gene Symbols NRP1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Neuropilin-1 Phe21-Pro856 Accession # P97333
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8829
Test Specificity Detects mouse Neuropilin-1 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant rat (rr) Neuropilin-1 is observed and no cross-reactivity with recombinant human (rh) Neuropilin-1, rhNeuropilin-2, or rrNeuropilin-2 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.