missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neuropilin-1 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5994A
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Neuropilin-1 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8829 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 761705 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P97333 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neuropilin-1 Phe21-Pro856 Accession # P97333 | |
| 100 Tests | |
| Primary | |
| Detects mouse Neuropilin-1 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant rat (rr) Neuropilin-1 is observed and no cross-reactivity with recombinant human (rh) Neuropilin-1, rhNeuropilin-2, or rrNeuropilin-2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction