missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Olfactomedin-1/Noelin-1 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF4636
This item is not returnable.
View return policy
Description
Olfactomedin-1/Noelin-1 Polyclonal specifically detects Olfactomedin-1/Noelin-1 in Mouse samples. It is validated for Western Blot.
Specifications
| Olfactomedin-1/Noelin-1 | |
| Polyclonal | |
| Biotin | |
| NP_062371 | |
| OLFM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Olfactomedin-1/Noelin-1 Ser2-Arg481 Accession # NP_062371 | |
| 50 μg | |
| Primary | |
| Detects mouse Olfactomedin-1/Noelin-1 in Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AMY, AMYolfactomedin related ER localized protein, neuroblastoma protein, Neuronal olfactomedin-related ER localized protein, NOE1, NOE1noelin, NOEL1, NOELIN, Noelin-1, NOELIN1, OlfA, olfactomedin 1, olfactomedin-1, Olfactomedin1, OLFM1, pancortin 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10439 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction