missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteoadherin/OSAD Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3308
This item is not returnable.
View return policy
Description
Osteoadherin/OSAD/OMD Polyclonal specifically detects Osteoadherin/OSAD/OMD in Mouse samples. It is validated for Western Blot.
Specifications
| Osteoadherin/OSAD/OMD | |
| Polyclonal | |
| Biotin | |
| O35103 | |
| OMD | |
| Mouse myeloma cell line NS0-derived recombinant mouse Osteoadherin/OSAD Gln21-Ile423 Accession # O35103 | |
| 50 μg | |
| Primary | |
| Detects mouse Osteoadherin/OSAD in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human Osteoadherin is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OMD, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2C, SLRR2Costeoadherin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4958 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction