missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PD-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7738V-100UG
This item is not returnable.
View return policy
Description
PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| PD-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q02242 | |
| PDCD1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-1 Leu25-Gln167 Accession # Q02242 | |
| RUO | |
| 5133 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 766104 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse PD-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human PD-1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction