missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pentraxin 2/SAP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2558
This item is not returnable.
View return policy
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Mouse samples. It is validated for Western Blot.
Specifications
| Pentraxin 2/SAP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P12246 | |
| APCS | |
| Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 2/SAP Gln21-Asp224 Accession # P12246 | |
| 100 μg | |
| Primary | |
| Detects mouse Pentraxin 2/SAP in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant rat Pentraxin 2/SAP is observed. In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2/SAP is observed, but no cross-reactivity with rhPentraxin 2/SAP in direct ELISAs is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 300103 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 325 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction