Learn More
Mouse Pentraxin 2/SAP Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2558-SP
 View more versions of this product
Product Code. 18720938
  • € 184.00 / 25µg
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pentraxin 2/SAP
Monoclonal
LYOPH
Western Blot 1 ug/mL
P12246
APCS
Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 2/SAP Gln21-Asp224 Accession # P12246
25 μg
Primary
Detects mouse Pentraxin 2/SAP in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant rat Pentraxin 2/SAP is observed. In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2/SAP is observed, but no cross-reactivity with rhPentraxin 2/SAP in direct ELISAs is observed.
Mouse
Purified
Western Blot
300103
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
325
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.