missing translation for 'onlineSavingsMsg'
Learn More

Mouse Persephin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18162551
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18162551 500 μg 500µg
18742618 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18162551 Supplier R&D Systems Supplier No. MAB2479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Persephin Monoclonal specifically detects Persephin in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Persephin
Applications Western Blot
Classification Monoclonal
Clone 240839
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O70300
Gene Alias persephin, PSP, PSPN
Gene Symbols PSPN
Host Species Rat
Immunogen E. coli-derived recombinant mouse Persephin Ala61-Gly156 Accession # O70300
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5623
Test Specificity Detects mouse Persephin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) AgRP, rmArtemin, rmCripto, recombinant drosophila DPP, recombinant human DPP-6, rhGDNF, rhLAP, rmMIS, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.