missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PGLYRP2/PGRP-L Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4704U-100UG
This item is not returnable.
View return policy
Description
PGLYRP2/PGRP-L Monoclonal specifically detects PGLYRP2/PGRP-L in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| PGLYRP2/PGRP-L | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8VCS0 | |
| PGLYRP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L Ser23-Asn530 Accession # Q8VCS0 | |
| RUO | |
| 114770 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 439728 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPL, PGLYRPLHMFT0141, PGRP2, PGRPL, PGRP-L, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse PGLYRP2/PGRP-L in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) PGRP-S and rmPGRP-1 beta is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction